missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Goat anti-Mouse IgG (H+L) Secondary Antibody, Rhodamine, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-6786
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Dot Blot, Fluorophore-linked immunosorbent assay.
Specifikationer
| IgG | |
| Polyclonal | |
| Rhodamine | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Mouse IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Prepared from monospecific antiserum by immunoaffinity chromatography using Mouse IgG coupled to agarose beads. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti- Goat Serum, Mouse IgG and Mouse Serum. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, ELISA | |
| 2.0 mg/mL | |
| Western Blot 1:1000 - 1:10000, Flow Cytometry 1:500 - 1:2500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry-Paraffin 1:200 - 1:500, Dot Blot, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Mouse | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering