missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit Complement C3 Polyclonal Anitbody, FITC, MP Biomedicals
Brand: MP Biomedicals 0855654
Additional Details : Weight : 0.00200kg
Description
Host Species: Goat
Species Reactivity: Rabbit
Specifications
Complement C3 | |
FITC | |
Affinity Purified | |
Primary |
Polyclonal | |
Goat | |
2 mL | |
Rabbit |