missing translation for 'onlineSavingsMsg'
Läs mer

SH2D1A Goat anti-Human, Polyclonal Antibody, Abnova™

Produktkod. 16099967
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16099967 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16099967 Leverantör Abnova Leverantörsnummer PAB6819.100ug

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Goat polyclonal antibody raised against synthetic peptide of SH2D1A.

This gene encodes a protein that plays a major role in the bidirectional stimulation of T and B cells. This protein contains an SH2 domain and a short tail. It associates with the signaling lymphocyte-activation molecule, thereby acting as an inhibitor of this transmembrane protein by blocking the recruitment of the SH2-domain-containing signal-transduction molecule SHP-2 to its docking site. This protein can also bind to other related surface molecules that are expressed on activated T, B and NK cells, thereby modifying signal transduction pathways in these cells. Mutations in this gene cause lymphoproliferative syndrome X-linked type 1 or Duncan disease, a rare immunodeficiency characterized by extreme susceptibility to infection with Epstein-Barr virus, with symptoms including severe mononucleosis and malignant lymphoma. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: C-HKRYFRKIKN

Specifikationer

Antigen SH2D1A
Användningsområden ELISA, Western Blot
Klassificering Polyclonal
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Beskrivning Goat polyclonal antibody raised against synthetic peptide of SH2D1A.
Utspädning ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulering In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gen SH2D1A
Gene Alias DSHP/EBVS/FLJ18687/FLJ92177/IMD5/LYP/MTCP1/SAP/XLP/XLPD
Gensymboler SH2D1A
Värd art Goat
Immunogen A synthetic peptide corresponding to human SH2D1A.
Reningsmetod Antigen affinity purification
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 4068
Målarter Human
Innehåll och lagring Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.