Learn More
TAZ, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TAZ.
Brand: Abnova PAB6881.100ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a protein that is expressed at high levels in cardiac and skeletal muscle. Mutations in this gene have been associated with a number of clinical disorders including Barth syndrome, dilated cardiomyopathy (DCM), hypertrophic DCM, endocardial fibroelastosis, and left ventricular noncompaction (LVNC). Multiple transcript variants encoding different isoforms have been described. A long form and a short form of each of these isoforms is produced; the short form lacks a hydrophobic leader sequence and may exist as a cytoplasmic protein rather than being membrane-bound. Other alternatively spliced transcripts have been described but the full-length nature of all these transcripts is not known. [provided by RefSeq
Sequence: C-HLKTQAEQLHNHSpecifications
TAZ | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
TAZ | |
TAZ | |
A synthetic peptide corresponding to human TAZ. | |
100 μg | |
Primary | |
This antibody is expected to recognize isoforms 1to 4 (NP_000107.1; NP_851828.1; NP_851829.1; NP_851830.1, respectively). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of TAZ. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
BTHS/CMD3A/EFE/EFE2/FLJ27390/G4.5/LVNCX/Taz1/XAP-2 | |
Goat | |
Antigen affinity purification | |
RUO | |
6901 | |
Human | |
Liquid |
For Research Use Only