Learn More
TBP, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TBP.
Brand: Abnova PAB24569.100ug
Additional Details : Weight : 0.02000kg
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes TBP, the TATA-binding protein. A distinctive feature of TBP is a long string of glutamines in the N-terminal. This region of the protein modulates the DNA binding activity of the C terminus, and modulation of DNA binding affects the rate of transcription complex formation and initiation of transcription. Mutations that expand the number of CAG repeats encoding this polyglutamine tract, and thus increase the length of the polyglutamine string, are associated with spinocerebellar ataxia 17, a neurodegenerative disorder classified as a polyglutamine disease. [provided by RefSeq
Sequence: C-TPQPIQNTNSLSSpecifications
TBP | |
Polyclonal | |
Goat polyclonal antibody raised against synthetic peptide of TBP. | |
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
GTF2D/GTF2D1/MGC117320/MGC126054/MGC126055/SCA17/TFIID | |
Goat | |
Antigen affinity purification | |
RUO | |
6908 | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
TBP | |
TBP | |
A synthetic peptide corresponding to amino acids 39-50 at internal region of human TBP. | |
100 μg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_003185.1; NP_001165556.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only