missing translation for 'onlineSavingsMsg'
Läs mer

VAV2, Goat, Polyclonal Antibody, Abnova™

Produktkod. 16049417
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16049417 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16049417 Leverantör Abnova Leverantörsnummer PAB6123.100ug

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Goat polyclonal antibody raised against synthetic peptide of VAV2.

VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: ETEAKYYRTLEDIEC

Specifications

Antigen VAV2
Användningsområden ELISA, Western Blot
Klassificering Polyclonal
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Beskrivning Goat polyclonal antibody raised against synthetic peptide of VAV2.
Utspädning ELISA (1:32000) Western Blot (1-1.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulering In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gen VAV2
Gensymboler VAV2
Värd art Goat
Immunogen A synthetic peptide corresponding to human VAV2.
Reningsmetod Antigen affinity purification
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 7410
Målarter Human
Innehåll och lagring Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.