Läs mer
GOLGA3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-09942-100UL
 Se fler versioner av denna produkt
Produktkod. 18332245
  • 4470.00 SEK / 100µL
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
GOLGA3 Polyclonal specifically detects GOLGA3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
GOLGA3
Polyclonal
Western Blot 1.0 ug/ml
GCP170Golgi peripheral membrane protein, golgi autoantigen, golgin subfamily a, 3, Golgi complex-associated protein of 170 kD, Golgi complex-associated protein of 170 kDa, Golgi membrane associated protein, golgin A3, Golgin subfamily A member 3, Golgin-160, golgin-165, male enhanced antigen-2, MEA-2, SY2/SY10 protein
The immunogen is a synthetic peptide directed towards the N-terminal region of Human GOLGA3. Peptide sequence VGSLGLPAHPREKKTSKSSKIRSLADYRTEDSNAGNSGGNVPAPDSTKGS
100 μg
Primary
Human
Purified
Western Blot
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
2802
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.