missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GOLGA7 Recombinant Protein
Brand: Abnova™ H00051125-P01.10ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH12032 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
40.81 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MRPQQAPVSGKVFIQRDYSSGTRCQFQTKFPAELENRIDRQQFEETVRTLNNLYAEAEKLGGQSYLEGCLACLTAYTIFLCMETHYEKVLKKVSKYIQEQNEKIYAPQGLLLTDPIERGLRVIEITIYEDRGMSSGR | |
GCP16/GOLGA3AP1/GOLGA7A/HSPC041/MGC21096/MGC4876 | |
GOLGA7 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
51125 | |
GOLGA7 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GOLGA7 | |
Human | |
Recombinant | |
Solution |