missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOSR2/Membrin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12095-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.Specifications
GOSR2/Membrin | |
Polyclonal | |
Unconjugated | |
GOSR2 | |
27 kDa Golgi SNARE protein, Bos1, GOSR2, GS27, Membrin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56494, 9570 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14653, O35166 | |
Gosr2 | |
GOSR2/Membrin Fusion Protein Ag2736 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |