Learn More
Corning™ Sterile Cutlery
Product Code. 11718623
Change view
Click to view available options
Product Type:
Knife
Spoon
Length (Metric):
125 mm
150 mm
175 mm
Quantity:
1000/Cs.
Unit Size:
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Length (Metric) Quantity Unit Size
11718623 Knife 175 mm 1000/Cs. Pack of 1000
11713114 Spoon 150 mm 1000/Cs. Pack of 1000
11847363 Spoon 125 mm 1000/Cs. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11718623 Supplier Corning™ Supplier No. COUT02
  • ONLINE EXCLUSIVE PRICE
    6400.00 SEK
    3331.05 SEK / Paket med 1000
    Save 3068.95 SEK
    47% Av
Beräknad leverans: 10-03-2027
Packaged individually in tamper evident zip bags. Gosselin™ Sterile Cutlery will help handle primary samples with limited cross contamination risks.
  • Resistant Polystyrene (PS) cutlery
  • Complete range
  • Sterile sample handling, single bag
TRUSTED_SUSTAINABILITY
Specifications
Product Type Knife
Material Polystyrene
Color White
Length (English) 6.8 in.
Length (Metric) 175 mm
Quantity 1000/Cs.
Sterility Sterile