missing translation for 'onlineSavingsMsg'
Learn More

GP73/GOLPH2 Mouse anti-Human, Clone: 3D1F1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66331-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16898105

  • 1656.73 SEK / 20µL
Estimated Shipment: 20-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi transmembrane protein. It processes protein synthesized in the rough endoplasmic reticulum and assists in the transport of protein cargo through the Golgi apparatus. The expression of this encoded protein has been observed to be upregulated in response to viral infection.
Specifications

Specifications

GP73/GOLPH2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NBJ4
GOLM1
GP73/GOLPH2 Fusion Protein Ag7207
20 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
3D1F1
Unconjugated
GOLM1
Golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2, GOLM1, GOLPH2, GP73
Mouse
Protein G
RUO
51280
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers