Learn More
GPALPP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566199
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: FGPALPPGFKK QDDSPPRPII GPALPPGFIK STQKSDKGRD DPGQQETDSS EDEDIIGPMP AKGPVNYNVT TEFEKRAQRM KE Highest antigen sequence identity to the following orthologs - mouse 82%, rat 79%.
GPALPP1 Polyclonal antibody specifically detects GPALPP1 in Human samples. It is validated for Immunocytochemistry.Specifications
GPALPP1 | |
Polyclonal | |
Unconjugated | |
GPALPP1 | |
1200011I18Rik; AD029; bA245H20.2; C6orf72; dJ12G14.2; GINM1; glycoprotein integral membrane 1; glycoprotein integral membrane protein 1; GPALPP motifs containing 1; GPALPP motifs-containing protein 1; Gpalpp1; hypothetical protein LOC532698; hypothetical protein LOC55425; Kiaa1704; lipopolysaccharide specific response-7 protein; lipopolysaccharide-specific response protein 7; LSR7; RGD1307254; UNQ710/PRO1361 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55425 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXQ4 | |
GPALPP1 | |
Recombinant Human GPALPP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only