missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPATCH8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568193
Additional Details : Weight : 0.01000kg
Description
GPATCH8 Polyclonal antibody specifically detects GPATCH8 in Human samples. It is validated for Western Blot.Specifications
GPATCH8 | |
Polyclonal | |
Unconjugated | |
GPATCH8 | |
5430405G24Rik; AU018890; ENSMUSG00000075516; Fbm1; Fyn-binding molecule 1; G patch domain containing 8; G patch domain-containing protein 8; Gpatc8; G-patch domain containing 8; GPATCH8; KIAA0553; mKIAA0553 | |
Rabbit | |
affinity chromatography | |
RUO | |
23131 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UKJ3 | |
GPATCH8 | |
Synthesized peptide derived from human GPATCH8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |