Learn More
GPBP1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17375-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
GPBP1L1 is a possible transcription factor.Specifications
GPBP1L1 | |
Polyclonal | |
Unconjugated | |
GPBP1L1 | |
GPBP1L1, RP11 767N6.1, SP192, Vasculin like protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313519, 60313, 77110 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3KR53, Q6NZP2, Q9HC44 | |
Gpbp1l1 | |
GPBP1L1 Fusion Protein Ag11180 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |