Läs mer
GPER/GPR30 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-31239
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
GPER/GPR30 Polyclonal specifically detects GPER/GPR30 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifikationer
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| Pig | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrigering av produktinnehåll
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering