missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR101 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550971
Additional Details : Weight : 0.01000kg
Description
GPR101 Polyclonal Antibody detects endogenous levels of GPR101 protein.
GPR101 Polyclonal antibody specifically detects GPR101 in Human, Mouse, Non-human Primate samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
GPR101 | |
Polyclonal | |
Unconjugated | |
GPR101 | |
G protein-coupled receptor 101; G protein-coupled receptor GPR101; Gm365; GPCR6; Gpr101; PAGH2; Probable G-protein coupled receptor 101; RGD1564196 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
245424, 83550 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q80T62, Q96P66 | |
GPR101 | |
Synthesized peptide derived from the C-terminal region of human GPR101. | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |