missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ GPR107 Monoclonal Antibody (1040717)
Beskrivning
Reconstitute at 0.5 mg/mL in sterile PBS.
GPR107 also named as KIAA1624 or LUSTR1 is 600 amino acid protein, which belongs to the LU7TM family. GPR107 localizes in membrane and is a multi-pass membrane protein. GPR107 is a promising candidate receptor for neuronostatin, and neuronostatin, interacting with GPR107, may play an important role in the central control of cardiovascular function.
Specifikationer
Specifikationer
| Antigen | GPR107 |
| Användningsområden | Western Blot |
| Klassificering | Monoclonal |
| Klona | 1040717 |
| Koncentration | 0.5 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with trehalose and no preservative |
| Gen | Gpr107 |
| Genaccessionsnr. | Q5VW38 |
| Gene Alias | 9930033D15Rik; AI790205; bA138E2.2; C530034M11; G protein-coupled receptor 107; GCDRP; Gpr107; KIAA1624; Lung seven transmembrane receptor 1; LUSTR1; mKIAA1624; Protein GPR107 |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?