missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR132 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17026-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
GPCR G2A is a G-protein linked receptor for lysophosphatidylcholine (LPC). It may play a role as a sensor of LPC levels at sites of inflammation to limit tissue-infiltrating-cells and progression to overt autoimmune disease and functions at the G2/M checkpoint to delay mitosis. It also may serve as a mechanism for T and B cells, and other cell types, to slow their proliferation and repair damaged DNA. It is highly expressed in macrophages and hematopoietic tissues rich in lymphocytes, like spleen and thymus, but weakly expressed in heart and lung. In atherosclerotic plaques, expression was observed around the lipid core and at the shoulder region. It is induced by DNA-damaging agents.Specifications
GPR132 | |
Polyclonal | |
Unconjugated | |
GPR132 | |
G protein coupled receptor 132, G2 accumulation protein, G2A, GPR132 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29933, 314480, 56696 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UNW8, Q9Z282 | |
Gpr132 | |
GPR132 Fusion Protein Ag10325 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |