missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR171 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GPR171 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839834
|
Proteintech
20307-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16829834
|
Proteintech
20307-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GPR171 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
O14626 | |
29909 | |
GPR171 Fusion Protein Ag14141 | |
Primary | |
-20°C | |
GPR171 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
G protein coupled receptor 171, GPR171, H963 | |
GPR171 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |