missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ GPR20 Recombinant Human Monoclonal Antibody (2C1)
GREENER_CHOICE

Produktkod. 30534435
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30534435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30534435 Leverantör Invitrogen™ Leverantörsnummer MA560829

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Human Recombinant Monoclonal Antibody

GPR20 Recombinant Monoclonal Antibody for ELISA

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. GPR signaling is an evolutionarily ancient mechanism used by all eukaryotes to sense environmental stimuli and mediate cell-cell communication. G protein-coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR20 is a 358 amino acid membrane protein that constitutively activates G(i) proteins without ligand stimulation. Also, GPR20 may be involved in the control of intracellular cAMP levels and mitogenic signaling. Interestingly, GPR20 is expressed in liver and certain regions of the brain, including putamen, caudate and thalamus, but is not expressed in hypothalamus, pons and frontal cortex.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen GPR20
Användningsområden ELISA
Klassificering Recombinant Monoclonal
Klona 2C1
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gen GPR20
Genaccessionsnr. Q99678
Gene Alias A430106B11Rik; CTD-3064M3.3; G protein-coupled receptor 20; G protein-coupled receptor 5-1; G protein-coupled receptor GPR20; Gpcr5-1; GPR20; G-protein coupled receptor 20; P2Y4
Gensymboler GPR20
Värd art Human
Immunogen Recombinant Human GPR20 protein.
Reningsmetod Affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 2843
Målarter Human, Monkey
Innehåll och lagring -20°C or -80°C if preferred
Produkttyp Antibody
Form Liquid
Isotyp IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.