missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ GPR27 Polyclonal Antibody

Beskrivning
Antibody detects endogenous levels of total GPR27.
G protein coupled receptors (GPCRs) are a large family of receptors that have a similar structure characterized by 7 transmembrane domains. Activation of GPCRs by extracellular stimuli such as neurotransmitters, hormones, or light induces an intracellular signaling cascade mediated by heterotrimeric GTP-binding proteins, or G proteins. The 375 amino acid GPCR GPR27 protein contains several features of the GPCR family, including 7 transmembrane domains, potential N linked glycosylation sites in the N terminal extracellular domain, and conserved cysteine residues, which may form a disulfide bridge, in the first and second extracellular loops.
Specifikationer
Specifikationer
| Antigen | GPR27 |
| Användningsområden | Western Blot, Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | GPR27 |
| Genaccessionsnr. | O54897, Q9JJH3, Q9NS67 |
| Gene Alias | G protein-coupled receptor 27; GPR27; Probable G-protein coupled receptor 27; SREB1; Super conserved receptor expressed in brain 1 |
| Gensymboler | GPR27 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?