Läs mer
Thermo Scientific™ GPS Series Lab Freezers
Protect routine samples with these freezers, in capacities ranging from a space-saving 265 liters to a spacious 1,048 liters.
Specifikationer
| Typ | General Purpose Freezer |
|---|---|
| Temperaturområde | -12°C to -25°C |
| Skåpsmaterial | Anti-corrosion Steel |
| Certifikat och efterlevnad | CE |
| Kontrolltyp | Digital |
Beskrivning
Protect routine samples with Thermo Scientific™ GPS Series Lab Freezers. With capacities ranging from a space-saving 265 liters to a spacious 1,048 liters, these freezers meet the demands of today's laboratories.
All models contain sample protection features designed for the rigorous demands of the laboratory environment.
- Integrated controller
- Digital display
- High- and low-temperature alarms
- Standard door locks
- Standard casters
- Automatic defrost
- Access ports
- CE marked
Recommended for:
General-purpose lab storage
Ordering Information:
Available in 50Hz regions only; not available in North America or other 60Hz regions.
Specifikationer
| General Purpose Freezer | |
| Anti-corrosion Steel | |
| Digital | |
| Warranty varies by country, please contact us for details | |
| Alarm | |
| 230 V | |
| 50 Hz |
| -12°C to -25°C | |
| CE | |
| Casters | |
| automatic | |
| EU | |
| 230 V 50 Hz | |
| R134A |