missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPT/ALT1 Mouse anti-Human, Unconjugated, Clone: 5F6B5, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GPT/ALT1 |
---|---|
Clone | 5F6B5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840216
|
Proteintech
67531-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16830216
|
Proteintech
67531-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The GPT/ALT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GPT/ALT1. This antibody recognizes human, pig antigen. The GPT/ALT1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
GPT/ALT1 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
2875 | |
Primary |
5F6B5 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Pig | |
IgG1 |