missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ GPX7 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518546
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.
Specifikationer
| GPX7 | |
| Polyclonal | |
| Unconjugated | |
| GPX7 | |
| 3110050F08Rik; AI327032; CL683; FLJ14777; glutathione peroxidase 6; glutathione peroxidase 7; GPX6; Gpx7; GPx-7; GSHPx-7; HGNC:4559; non-selenocysteine containing phospholipid hydroperoxide glutathione peroxidase; NPGPx; UNQ469/PRO828 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2882 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96SL4 | |
| GPX7 | |
| Synthetic peptide sequence (CNQFGQQEPDSNK) corresponding to the internal amino acids of GPX7 (aa 86-99). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering