missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), Alexa Fluor™ 647, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen RM3021
Additional Details : Weight : 0.50000kg
Description
Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.Specifications
Ly-6G/Ly-6C | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P0CW02, P0CW03, P35461 | |
Ly6c1, Ly6c2, Ly6g | |
Normal murine bone marrow cells | |
1 mL | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry | |
RB6-8C5 | |
Alexa Fluor 647 | |
Ly6c1 | |
AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN | |
Rat | |
purified | |
RUO | |
100041546, 17067, 546644 | |
4° C | |
Liquid |