Läs mer
Tocris Bioscience™ GR 127935 hydrochloride
GREENER_CHOICE
Produktkod. 30001260
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001260 50 mg
30001259 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001260 Leverantör Tocris Bioscience™ Leverantörsnummer 1477/50
Potent and selective 5-HT1B and 5-HT1D antagonist
Potent and selective 5-HT1B/1D receptor antagonist (pKi values are 8.5 for both guinea pig 5-HT1D and rat 5-HT1B receptors). Displays > 100-fold selectivity over 5HT1A, 5-HT2A, 5-HT2C receptors and other receptor types. Centrally active following oral administration.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 148642-42-6
Molekylformel C29H32ClN5O3
Specifikationer
Kemiskt namn eller material GR 127935 hydrochloride
Kvantitet 50 mg
Formel vikt 534.06
Renhet >99%
Molekylvikt (g/mol) 534.057