missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ GR 203040

Description
GR 203040 is a potent and selective NK1 receptor antagonist. Potently inhibits substance P binding to NK1 receptors (pKi = 10.1 to 10.5). Displays antiemetic activity in vivo.
Specifications
Specifications
| CAS | 168398-02-5 |
| Synonym | (2S,3S)-N-[[2-Methoxy-5-(1H-tetrazol-1-yl)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride |
| Kvantitet | 25 mg |
| Molekylformel | C20H24N6O.2HCl |
| Formel vikt | 437.37 |
| Renhet | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?