Läs mer
GR 203040, Tocris Bioscience™
GREENER_CHOICE
Potent, selective NK1 receptor antagonist
Brand:  Tocris Bioscience 4647/25
Encompass
Produktkod. 15907900
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
GR 203040 is a potent and selective NK1 receptor antagonist. Potently inhibits substance P binding to NK1 receptors (pKi = 10.1 to 10.5). Displays antiemetic activity in vivo.
Specifikationer
Specifikationer
168398-02-5
(2S,3S)-N-[[2-Methoxy-5-(1H-tetrazol-1-yl)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride
25 mg
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer