missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Munktell Filtrak™ Grade 00H Quantitative Ashless Filter Papers
Recommended for use where a high purity is required for quantitative analysis.
Brand: Munktell Filtrak™ 100004
Additional Details : Weight : 0.10800kg
Description
- Made from pure cellulose
- Ash content of ashless grades is less than 0.01
- Basis Weight: 80g/m2
Specifications
Disc | |
1 to 2 μm | |
00H |
Cellulose | |
160s/10 mL | |
125 mm |