missing translation for 'onlineSavingsMsg'
Learn More

Munktell Filtrak™ Grade 00H Quantitative Ashless Filter Papers

Recommended for use where a high purity is required for quantitative analysis.

Brand:  Munktell Filtrak™ 100008

Additional Details : Weight : 0.68400kg

 View more versions of this product

Product Code. 11382954

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Made from pure cellulose
  • Ash content of ashless grades is less than 0.01
  • Basis Weight: 80g/m2
Specifications

Specifications

Disc
1 to 2 μm
00H
Cellulose
160s/10 mL
320 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions