Learn More
Munktell Filtrak™ Grade 00R Quantitative Ashless Filter Papers
Recommended for use where a high purity is required for quantitative analysis.
Brand:  Munktell Filtrak™ 100058
Additional Details : Weight : 0.09000kg
Product Code. 11372994
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Made from pure cellulose
  • Ash content of ashless grades is less than 0.01
  • Basis Weight: 80g/m2
Specifications
Specifications
Disc
Cellulose
Ashless
110 mm