missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 10 Smooth Surface Filter Discs Qualitative Filter Papers

Used for routine analyses like clarification, determination of substances, discs with center hole for technical applications

Brand:  Sartorius™ FT-3-352-125

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11780486

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Made of refined pulp and cotton linters with an >95% α-cellulose content
  • Ash content between 0.1–0.15% (grade 100/N <0.1%)
  • Wet-strengthened
  • Supplied in rolls, sheets, discs and folded filters as well as customer-specific cuts
Specifications

Specifications

Disc
7.5μm/sec.
10
0.33 mm
125 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
50/Pk.
Qualitative Filter Paper
125 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers