missing translation for 'onlineSavingsMsg'
Learn More

Munktell Filtrak™ Grade 1003 General Purpose Qualitative Filter Paper

Suitable for typical general filtration tasks.

Brand:  Munktell Filtrak™ 120031

Additional Details : Weight : 0.05940kg

Product Code. 11373124

  • 115.00 SEK / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

  • Made from pulp with a high alpha-cellulose content and an average ash content of 0.1%
  • Basis Weight: 90g/m2
Specifications

Specifications

Disc
12 to 15 μm
1003
Cellulose
10s/10 mL
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers