Learn More
Munktell Filtrak™ Grade 1600 Pulp Testing Papers
Grade 1600 Test Paper is often used when preparing laboratory sheets of most forms of pulp for physical testing in accordance with ISO Test Method 5269-1: 2005, Part 1.
967.00 SEK - 2455.00 SEK
Specifications
Basis Weight | 250 ± 25 g/m2 |
---|---|
Format | Disc |
For Use With (Application) | Physical test prep |
Grade | Grade 1600 |
Quantity | 200/Pk. |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11317254
|
Munktell Filtrak™
143039 |
140 mm |
1010.00 SEK
Pack of 200 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11307254
|
Munktell Filtrak™
143037 |
125 mm |
967.00 SEK
Pack of 200 |
In Stock
Log in to see stock. |
|||||
11367254
|
Munktell Filtrak™
143042 |
220 mm |
2455.00 SEK
Pack of 200 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
Description
- In circle disc format
- Available in a range of diameters
- Klemm absorbency of 70 ± 20 mm
- Dimensional change of less than 3%
- Water uptake of 450 ± 50 g/m²
- Provided 200 per pack
Specifications
250 ± 25 g/m2 | |
Physical test prep | |
200/Pk. |
Disc | |
Grade 1600 | |
Pulp Test Paper |