missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Munktell Filtrak™ Grade 3 Qualitative High Purity Lab Filter Papers
For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis.
Brand: Munktell Filtrak™ 110065
Additional Details : Weight : 0.06300kg
Description
- Made of pure cotton linters
- For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si-determination in steel and pig iron analysis
- Basis Weight: 90g/m2
Specifications
Disc | |
<10 μm | |
3 |
Cotton Linters | |
11s/10 mL | |
90 mm |