Läs mer
Granisetron-d3, TRC
Produktkod. 30240842
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240842 1 mg
30240663 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240842 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG7800021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1224925-64-7
Molekylformel C18H21D3N4O
Molekylvikt (g/mol) 315.43
Synonym 1-Methyl-N-[(3-endo)-9-(methyl-d3)-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide, BRL-43694A-d3, Kytril-d3;
LEDER CN1C2=C(C=CC=C2)C(C(N[C@@H]3C[C@@H](N4C([2H])([2H])[2H])CCC[C@@H]4C3)=O)=N1
Specifikationer
Kemiskt namn eller material Granisetron-d3
Kvantitet 1 mg
InChI formel InChI=1S/C18H24N4O/c1-21-13-6-5-7-14(21)11-12(10-13)19-18(23)17-15-8-3-4-9-16(15)22(2)20-17/h3-4,8-9,12-14H,5-7,10-11H2,1-2H3,(H,19,23)
Formel vikt 315.2138