Learn More
Granzyme A Monoclonal Antibody (CB9), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1394.00 SEK - 2670.00 SEK
Specifications
Antigen | Granzyme A |
---|---|
Clone | CB9 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | eFluor 660 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15560087
|
Affymetrix eBioscience
50-9177-42 |
100 Tests |
2670.00 SEK
100 tests |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15550087
|
Affymetrix eBioscience
50-9177-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Cytolytic T lymphocytes (CTL) and natural killer (NK) cells share the remarkable ability to recognize, bind, and lyse specific target cells. They are thought to protect their host by lysing cells bearing on their surface 'nonself' antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein described here is a T cell- and natural killer cell-specific serine protease that may function as a common component necessary for lysis of target cells by cytotoxic T lymphocytes and natural killer cells.
Granzyme A Monoclonal antibody specifically detects Granzyme A in Human samples. It is validated for Flow Cytometry.Specifications
Granzyme A | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Human | |
3001 | |
IgG1 κ | |
Affinity chromatography | |
GZMA |
CB9 | |
Monoclonal | |
Liquid | |
RUO | |
P12544 | |
GZMA | |
Primary | |
Antibody |
For Research Use Only.