missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Granzyme B Monoclonal Antibody (GB11), PE, eBioscience™
GREENER_CHOICE

Produktkod. 15537146
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 Tests
Förpackningsstorlek:
25 tester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15537146 25 Tests 25 tester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15537146 Leverantör Invitrogen™ Leverantörsnummer 12889941

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

Description: The GB11 antibody reacts with human Granzyme B (GrB). GrB is one of the family of serine proteases, known as granzymes, that is located in the granules of cytotoxic T cells and NK cells. Granzyme B has been described also as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation. GrB has been called CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. GrB is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved has been identified as mannose 6-phosphate receptor. This receptor functions as a death receptor for granzyme B during cytotoxic T cell-induced apoptosis. Applications Reported: The GB11 antibody has been reported useful for intracellular staining followed by flow cytometric analysis. Applications Tested: This GB11 antibody has been pre-titrated and tested on by intracellular staining and flow cytometric analysis of both cultured normal human peripheral blood cells and mouse splenocytes. This can be used at test size: 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; L...

Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Granzyme B
Användningsområden Flow Cytometry
Klassificering Monoclonal
Klona GB11
Koncentration 5 μL/Test
Konjugera PE
Formulering PBS with BSA and 0.09% sodium azide; pH 7.2
Gen GZMB
Genaccessionsnr. P10144
Gene Alias AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3E
Gensymboler GZMB
Värd art Mouse
Reningsmetod Affinity chromatography
Kvantitet 25 Tests
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 3002
Målarter Human
Innehåll och lagring 4°C, store in dark, DO NOT FREEZE!
Produkttyp Antibody
Form Liquid
Isotyp IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.