Läs mer
Invitrogen™ Granzyme K Monoclonal Antibody (G3H69), PerCP-eFluor™ 710, eBioscience™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 46-8897-42
Description
Description: This G3H69 monoclonal antibody reacts with human Granzyme K. Granzyme K is one of five granzyme serine proteases that have been identified in humans. These proteins are expressed in the granules of NK cells and cytotoxic T cells, and are critical for the induction of target cell apoptosis through the cleavage of intracellular substrates. Granzyme A and Granzyme K are both tryptases and appear to show overlapping, though not identical, substrate specificity. This functional similarity is believed to account for the minimal decrease in cytotoxicity of Granzyme A-deficient CTLs. Applications Reported: This G3H69 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This G3H69 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood monocytes. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Sam...
Granzyme K (GrK) belongs to a family of trypsin-like serine proteases localised in the cytotoxic granules of activated T cells and NK cells. It encodes a 28 kDa serine protease whose gene is located on chromosome 5q11-12 close to the granzyme A-encoding gene. Like granzyme A, it has a trypsin-like specifity cleaving at the basic residues arginine and lysine. Granzyme K triggers rapid cell death independently of caspase activation with single-stranded DNA nicks and is primarily expressed in thymus, lung, spleen and peripheral blood leukocytes.
Specifications
| Granzyme K | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P49863 | |
| GZMK | |
| Affinity chromatography | |
| RUO | |
| 3003 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| Responsibly packaged |
| Flow Cytometry | |
| G3H69 | |
| PerCP-eFluor 710 | |
| GZMK | |
| Fragmentin-3; granzyme 3; granzyme K; granzyme K (granzyme 3; tryptase II); granzyme K (serine protease, granzyme 3; tryptase II); granzyme-3; GrK; GZMK; NK-Tryp-2; NK-tryptase-2; RNK-Tryp-2; TRYP2; tryptase II | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.