missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme M Monoclonal Antibody (4B2G4), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
2040.00 SEK - 4870.00 SEK
Specifications
Antigen | Granzyme M |
---|---|
Clone | 4B2G4 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16606284
|
Affymetrix eBioscience
12-9774-41 |
25 Tests |
2040.00 SEK
25 tests |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16616284
|
Affymetrix eBioscience
12-9774-42 |
100 Tests |
4870.00 SEK
100 tests |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cleaves peptide substrates after methionine, leucine, and norleucine. Physiological substrates include EZR, alpha- tubulins and the apoptosis inhibitor BIRC5/Survivin. Promotes caspase activation and subsequent apoptosis of target cells.Specifications
Granzyme M | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
granzyme M; granzyme M (lymphocyte met-ase 1); GZMM; Hu-Met-1; Lmet1; lymphoctye Met-ase 1; lymphocyte met-ase 1; MET1; Met-1 serine protease; Met-ase; MMET-1; natural killer cell granular protease; RNK-Met-1 | |
GZMM | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
GZMM |
4B2G4 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
P51124 | |
3004 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |