Learn More
Graphite Fusion Crucible Lid, Thermo Scientific Chemicals
Product Code. 15473875
Change view
Click to view available options
Diameter (Metric) Outer:
2.54cm
3.2cm
Quantity:
15/Pk
15/Pk.
Description:
For stock No. 14747
For stock No. 40794
Unit Size:
Pack of 15
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Quantity Description Unit Size
15473875 2.54cm 15/Pk For stock No. 40794 Pack of 15
15413895 3.2cm 15/Pk. For stock No. 14747 Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15473875 Supplier Thermo Scientific Alfa Aesar Supplier No. 042315.UA
 Restricted Item
I lager
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Graphite is used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description For stock No. 40794
Material Graphite
Quantity 15/Pk
Diameter (Metric) Outer 2.54cm
Product Type Graphite Fusion Crucible Lid