Learn More
Motic™ Green Filter, BA210, BA310 and BA410
Suitable for BA210, BA310 and BA410 upright compound microscopes. Motic™ Green Filter, BA210, BA310 and BA410 improves achromat/planachromat objective performance and phase contrast image quality.
Brand: Motic™ 1101000300312
Additional Details : Weight : 0.01000kg
Description
- Improves achromat and planachromat objective performance.
- Produces the highest quality phase contrast images.
Specifications
Green | |
OD: 45mm | |
BA210, BA310 and BA410 Microscopes |