Learn More
Motic™ Green Filter, BA210, BA310 and BA410
Suitable for BA210, BA310 and BA410 upright compound microscopes. Motic™ Green Filter, BA210, BA310 and BA410 improves achromat/planachromat objective performance and phase contrast image quality.
Brand:  Motic™ 1101000300312
Additional Details : Weight : 0.01000kg
Product Code. 15495152
  • 377.00 SEK / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
Description
Description
  • Improves achromat and planachromat objective performance.
  • Produces the highest quality phase contrast images.
Specifications
Specifications
Green
OD: 45mm
BA210, BA310 and BA410 Microscopes