Learn More
GRIA1 Mouse anti-Human, Unconjugated, Clone: 1F4A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67642-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The GRIA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GRIA1. This antibody recognizes human, pig, mouse, rat, rabbit antigen. The GRIA1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
GRIA1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig, Rabbit |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1F4A1 | |
Unconjugated | |
20 μL | |
2890 | |
IgG2b |