Learn More
GRIA1 Mouse anti-Human, Unconjugated, Clone: 1F4A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67642-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840386
  • 1656.73 SEK / 20µL
Estimated Shipment: 26-04-2024
to see stock.
Description
Description
The GRIA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GRIA1. This antibody recognizes human, pig, mouse, rat, rabbit antigen. The GRIA1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
GRIA1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
Western Blot, Immunohistochemistry, Immunofluorescence
1F4A1
Unconjugated
20 μL
2890
IgG2b