Learn More
GRID2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563583
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AVYEHVRMKG LNPFERDSMY SQMWRMINRS NGSENNVLES QAGIQKVKYG NYAFVWDAAV LEYVAINDPD CSFYTIGNTV ADR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
GRID2 Polyclonal antibody specifically detects GRID2 in Human samples. It is validated for Immunohistochemistry.Specifications
GRID2 | |
Polyclonal | |
Unconjugated | |
GRID2 | |
B230104L07Rik; cpr; creeper; GluD2; gluR delta-2 subunit; GLURD2; GluRdelta2; glutamate ionotropic receptor delta type subunit 2; glutamate receptor; glutamate receptor delta-2 subunit; glutamate receptor ionotropic, delta-2; glutamate receptor, ionotropic, delta 2; Grid2; ho; hotfoot; Lc; Lc<J>; lurcher; minisatellite 10ac detected by probe MMS10; MMS10-AC; Ms10ac; nmf408; SCAR18; tpr | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2895 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43424 | |
GRID2 | |
Recombinant protein corresponding to Human GRID2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only