Läs mer
Grifolic Acid, TRC
Produktkod. 17964635
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17964635 5 mg
17912085 1 mg
17900635 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17964635 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG7865005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80557-12-6
Molekylformel C23 H32 O4
Molekylvikt (g/mol) 372.4978
Synonym 2,4-Dihydroxy-6-methyl-3-[(2E,6E)-3,7,11-trimethyl-2,6,10-dodecatrien-1-yl]benzoic Acid, (E,E)-2,4-dihydroxy-6-methyl-3-(3,7,11-trimethyl-2,6,10-dodecatrienyl)benzoic Acid, 2,4-Dihydroxy-6-methyl-3-[(2E,6E)-3,7,11-trimethyl-2,6,10-dodecatrienyl]benzoic Acid, Ilicicolinic Acid B
IUPAC-namn 2,4-dihydroxy-6-methyl-3-[(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl]benzoic acid
LEDER CC(=CCC\C(=C\CC\C(=C\Cc1c(O)cc(C)c(C(=O)O)c1O)\C)\C)C
Specifikationer
Kemiskt namn eller material Grifolic Acid
Kvantitet 5 mg
InChI formel InChI=1S/C23H32O4/c1-15(2)8-6-9-16(3)10-7-11-17(4)12-13-19-20(24)14-18(5)21(22(19)25)23(26)27/h8,10,12,14,24-25H,6-7,9,11,13H2,1-5H3,(H,26,27)/b16-10+,17-12+
Formel vikt 372.2301