missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GRIK2 (Human) Recombinant Protein
Human GRIK2 full-length ORF ( AAH37954, 1 a.a. - 353 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH37954 |
---|---|
Gene ID (Entrez) | 2898 |
Name | glutamate receptor, ionotropic, kainate 2 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115761
|
Abnova™
H00002898-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16125761
|
Abnova™
H00002898-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MKIIFPILSNPVFRRTVKLLLCLLWIGYSQGTTHVLRFGGIFEYVESGPMGAEELAFRFAVNTINRNRTLLPNTTLTYDTQKINLYDSFEASKKACDQLSLGVAAIFGPSHSSSANAVQSICNALGVPHIQTRWKHQVSDNKDSFYVSLYPDFSSLSRAILDLVQFFKWKTVTVVYDDSTGLIRLQELIKAPSRYNLRLKIRQLPADTKDAKPLLKEMKRGKEFHVIFDCSHEMAAGILKQALAMGMMTEYYHYIFTTLDLFALDVEPYRYSGVNMTGFRILNTENTQVSSIIEKWSMERLQAPPKPDSGLLDGFMTMEFHSCCPGWSAMARSHLTAASASWVQAILLPQPPE
Specifications
AAH37954 | |
glutamate receptor, ionotropic, kainate 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
EAA4, GLR6, GLUK6, GLUR6, MGC74427, MRT6 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
2898 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GRIK2 | |
GST |