missing translation for 'onlineSavingsMsg'
Learn More

GRIN2B Mouse anti-Human, Mouse, Rat, Clone: 1C5E12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66565-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878555

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer′s Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal domit 6-related mental retardation.
Specifications

Specifications

GRIN2B
1C5E12
Unconjugated
GRIN2B
Liquid
GRIN2B
GRIN2B Fusion Protein Ag16718
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Western Blot
0.66 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q00960, Q01097, Q13224
GluN2B, GRIN2B, hNR3, NMDAR2B, NR2B, NR3
Mouse
IgG1
150 μL
-20°C
Primary
14812, 24410, 2904
Videos
SDS
Documents
Certificates
Promotions

Promotions