missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ GRK3 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 30534326
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30534326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30534326 Leverantör Invitrogen™ Leverantörsnummer PA5148902

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

ADRBK2, or adrenergic beta receptor kinase 2 is a serine/threonine kinase that is thought to act as a regulator of receptor function. Overall, the ADRBK2 enzyme, also known as GRK3, has 85% amino acid similarity with ADRBK1, with the protein kinase catalytic domain having 95% similarity. The ADRBK2 mRNA is approximately 8 kilobases with a distribution similar to that of ADRBK1. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen GRK3
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 0.2 mg/mL
Konjugera Unconjugated
Formulering PBS with trehalose and no preservative
Gen GRK3
Genaccessionsnr. P35626
Gene Alias 4833444A01Rik; ADRBK2; Adrbk-2; adrenergic receptor kinase, beta 2; adrenergic receptor kinase, beta 2 (G-protein-linked receptor kinase); adrenergic, beta, receptor kinase 2; AI851927; AW551196; BARK2; Bark-2; beta ARK2; beta-adrenergic receptor kinase 2; Beta-ARK-2; G protein-coupled receptor kinase 3; G-protein-coupled receptor kinase 3; Grk3
Gensymboler GRK3
Värd art Sheep
Immunogen E. coli-derived recombinant human GRK3, Pro469-Leu688.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 157
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Lyophilized
Isotyp IgG
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.