missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK6 Recombinant Human Protein
GRK6 Recombinant Human Protein
Brand: Invitrogen PV3661
Additional Details : Weight : 0.05250kg
Description
GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine⁄threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation.Specifications
NP_001004106 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
94kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
GRK6, GPRK6 | |
GRK6 was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 5mM MgCl2, 0.25mM EGTA, 0.1mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 240μg/mL partially dephosphorylated casein substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
GRK6 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.51mg/mL | |
2870, 3553 | |
GRK6 Recombinant Human Protein | |
–80°C | |
1 Each | |
GRK6 Recombinant Human Protein | |
GRK | |
29.9nmoles of phosphate transferred to partially dephosphorylated casein substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.