Läs mer
Thermo Scientific™ Growth Hormone Releasing Peptide-2
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
25 mg
Förpackningsstorlek:
25mg
5mg
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
Specifikationer
| CAS | 158861-67-7 |
| Innehåll och lagring | Store at -20°C |
| Formatera | Solid |
| Kvantitet | 5 mg |